Aluminium hydroxide ≥99%, dried gel
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1317-500G
93-1317-2000G
100203-378EA
84.92
USD
100203-378
100198-302
Aluminium hydroxide ≥99%, dried gel
Aluminium hydroxide
Formula:
Al(OH)₃ Melting Pt: 300 °C Density: 2.42 |
MDL Number:
MFCD00003420 CAS Number: 21645-51-2 EINECS: 244-492-7 Merck Index: 12,00355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...