Magnesium ≥99.8%, rods

Supplier: STREM CHEMICALS INC MS
Warning

93-1295-1ROD 93-1295-5ROD
100200-050EA 67.94 USD
100200-050 100210-172
Magnesium ≥99.8%, rods
Magnesium
Formula: Mg
Boiling Pt: 1107 °C
Melting Pt: 657 °C
Density: 1.74
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR