Magnesium acetylacetonate, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Magnesium bis(2,4-pentanedionato-O,O'), Bis(2,4-pentanedionato-O,O')magnesium, Magnesiumacetylacetonate, Magnesium 2,4-pentanedionate

93-1280-25G 93-1280-100G
100204-164EA 249.11 USD
100204-164 100203-368
Magnesium acetylacetonate, anhydrous ≥98%
Magnesium acetylacetonate
Formula: C₁₀H₁₄MgO₄
Melting Pt: Dec.
MDL Number: MFCD00013497
CAS Number: 14024-56-7
EINECS: 237-857-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR