Magnesium nitrate hexahydrate ≥99.999% (Mg basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-1237-25G 93-1237-100G
100211-206EA 202.4 USD
100211-206 100200-048
Magnesium nitrate hexahydrate ≥99.999% (Mg basis), PURATREM
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 89 °C
Density: 1.64
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 13,05697

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR