Sodium nitrate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1161-2000G
93-1161-500G
100210-162EA
229.29
USD
100210-162
100206-910
Sodium nitrate ≥99% ACS
Sodium nitrate
Formula:
NaNO₃ Boiling Pt: 380 °C (1013 hPa) Melting Pt: 306.8 °C Density: 2.261 Storage Temperature: Ambient |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III REACH: 01-2119488221-41 Merck Index: 13,08720 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...