Sodium iodide, anhydrous ≥99% (H2O < 1%)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

93-1156-100G 93-1156-500G
100203-364EA 107.57 USD
100203-364 100208-082
Sodium iodide, anhydrous ≥99% (H2O < 1%)
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.667
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR