Sodium hexafluorophosphate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1146-10G
93-1146-50G
100203-362EA
80.68
USD
100203-362
100200-040
Sodium hexafluorophosphate ≥99%
Sodium hexafluorophosphate
Formula:
F₆NaP MW: 167.95 g/mol Melting Pt: 450 °C Density: 3.375 |
MDL Number:
MFCD00011122 CAS Number: 21324-39-0 EINECS: 244-333-1 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...