Sodium azide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium triazide
93-1112-500G
93-1112-100G
100208-706EA
375.08
USD
100208-706
100205-270
Sodium azide ≥98%
Sodium azide
Formula:
NaN₃ MW: 65.01 g/mol Melting Pt: 300 °C (dec.) Density: 1.846 Storage Temperature: Ambient |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 EINECS: 247-852-1 UN: 1687 ADR: 6.1,II Merck Index: 13,08653 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...