EDTA tetrasodium salt dihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tetrasodium ethylenediaminetetraacetate dihydrate, Edetic acid tetrasodium salt dihydrate, Kestranal® 4S 2 H2O, Ethylenedinitrilotetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetra-acetic acid tetrasodium salt dihydrate, Ethylenediaminetetraacetic acid sodium hydrate, Tetrasodium Ethylenediaminetetraacetate Dihydrate, Kestranal® 4S dihydrate
93-1104-2000G
93-1104-500G
100203-356EA
373.66
USD
100203-356
100211-200
EDTA tetrasodium salt dihydrate ≥99%
EDTA tetrasodium salt dihydrate
Formula:
C₁₀H₁₂N₂Na₄O₈·2H₂O Melting Pt: > 300 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150026 CAS Number: 10378-23-1 EINECS: 200-573-9 Merck Index: 13,03544 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...