Sodium niobate ≥99.9% (Nb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Sodium niobium oxide
93-1057-5G
93-1057-25G
100197-654EA
65.11
USD
100197-654
100196-448
Sodium niobate ≥99.9% (Nb basis)
Sodium niobate
Formula:
NaNbO₃ Melting Pt: 1250 °C Density: 4,42 g/cm³ (25 °C) |
MDL Number:
MFCD00014230 CAS Number: 12034-09-2 EINECS: 234-798-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...