Sodium hexafluoroantimonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Sodium hexafluoroantimonate(V)

93-1006-100G 93-1006-25G
100198-286EA 220.8 USD
100198-286 100205-266
Sodium hexafluoroantimonate ≥99%
Sodium hexafluoroantimonate
Formula: F₆NaSb
Density: 3.375
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR