Cyanogen bromide ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

93-1001-100G 93-1001-25G
100211-198EA 256.18 USD
100211-198 100200-504
Cyanogen bromide ≥97%
Cyanogen bromide
Formula: CBrN
Boiling Pt: 61.4 °C
Melting Pt: 52 °C
Density: 2.015
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 12,02763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR