Carbon ≥99%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Charcoal
93-0601-1000G
93-0601-250G
100205-264EA
76.43
USD
100205-264
100197-650
Carbon ≥99%, powder
Carbon
Formula:
C MW: 12.01 g/mol Boiling Pt: 4200 °C Melting Pt: 3652…3697 °C (subl.) Density: 1.8…2.1 (amorphous) |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 Merck Index: 13,01818 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...