Triethyl borate ≥98.5%

Supplier: STREM CHEMICALS INC MS
Danger

93-0530-100G 93-0530-500G
100200-026EA 69.35 USD
100200-026 100200-028
Triethyl borate ≥98.5%
Triethyl borate
Formula: C₆H₁₅BO₃
Boiling Pt: 117.4 °C
Melting Pt: –84.8 °C
Density: 0.864
Flash Pt: 52 °F
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR