Beryllium ≥99%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0420-5G
93-0420-25G
100201-814EA
72.18
USD
100201-814
100204-664
Beryllium ≥99%, powder
Beryllium
Formula:
Be Boiling Pt: 2970 °C (5 mmHg) Melting Pt: 1278 °C Density: 1,85 |
MDL Number:
MFCD00134032 CAS Number: 7440-41-7 EINECS: 231-150-7 UN: 1567 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...