Lithium carbonate ≥99.99% (Li basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-0312-25G 93-0312-100G
100200-014EA 86.34 USD
100200-014 100209-398
Lithium carbonate ≥99.99% (Li basis), PURATREM
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Boiling Pt: 1310 °C (dec.)
Melting Pt: 720
Density: 2.11
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR