Bismuth(III) fluoride ≥99.99% (Bi basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
83-3000-10G
83-3000-50G
100204-114EA
107.57
USD
100204-114
100206-138
Bismuth(III) fluoride ≥99.99% (Bi basis), PURATREM
Bismuth (III) fluoride
Formula:
BiF₃ Boiling Pt: 230 °C (1013 hPa) Melting Pt: 727 °C Density: 5.32 |
MDL Number:
MFCD00010893 CAS Number: 7787-61-3 EINECS: 232-124-8 UN: 3260 ADR: 8,III Merck Index: 12,01305 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...