Lead(II) iodide ≥99.999% (Pb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

82-0750-50G 82-0750-10G
100198-922EA 67.94 USD
100198-922 100200-500
Lead(II) iodide ≥99.999% (Pb basis), PURATREM
Lead (II) iodide
Formula: I₂Pb
MW: 461.01 g/mol
Boiling Pt: 954 °C
Melting Pt: 402 °C
Density: 6.16
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 12,05430

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR