Thallium ≥99.9%, granules
Supplier: STREM CHEMICALS INC MS
|
Danger
|
81-0054-25G
81-0054-100G
100206-852EA
184
USD
100206-852
100197-176
Thallium ≥99.9%, granules
Thallium
Formula:
Tl Boiling Pt: 1457 °C Melting Pt: 301 °C Density: 11.85 Storage Temperature: Ambient |
MDL Number:
MFCD00134063 CAS Number: 7440-28-0 EINECS: 231-138-1 UN: 1707 ADR: 6.1,II Merck Index: 12,09391 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...