Dipotassium hexachloroiridate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium hexachloroiridate(IV), Potassium hexachloroiridate (IV)

77-6500-5G 77-6500-1G
100201-022EA 1360.18 USD
100201-022 100201-020
Dipotassium hexachloroiridate ≥99%
Dipotassium hexachloroiridate
Formula: Cl₆IrK₂
Melting Pt: Dec.
Density: 3,546
MDL Number: MFCD00011369
CAS Number: 16920-56-2
EINECS: 240-976-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR