Trifluoromethanesulfonic acid ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trifluoromethanesulphonic acid, Trifluoromethanesulfonic acid

09-7410-10G 09-7410-50G
100197-356EA 63.69 USD
100197-356 100210-970
Trifluoromethanesulfonic acid ≥99%
Trifluoromethanesulphonic acid

Organo Fluorine

Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C
Melting Pt: 35 °C
Density: 1,696
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR