Trifluoroacetic acid ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: TFA

09-7160-100G 09-7160-500G
100196-020EA 83.51 USD
100196-020 100206-276
Trifluoroacetic acid ≥99%
Trifluoroacetic acid

Organo Fluorine

Formula: C₂HF₃O₂
Boiling Pt: 74 °C
Melting Pt: –15.4 °C
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR