Tetrabutylammonium tetrafluoroborate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

09-6635-25G 09-6635-5G
100204-880EA 159.94 USD
100204-880 100198-570
Tetrabutylammonium tetrafluoroborate ≥99%
Tetrabutylammonium tetrafluoroborate

Organo Fluorine

Formula: C₁₆H₃₆BF₄N
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR