Pentafluoropropionic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid

09-5740-10G 09-5740-50G
100208-782EA 90.58 USD
100208-782 100203-714
Pentafluoropropionic acid ≥97%
Pentafluoropropionic acid

Organo Fluorine

Formula: C₃HF₅O₂
Boiling Pt: 96…98 °C
Density: 1,561
Storage Temperature: Ambient
MDL Number: MFCD00004170
CAS Number: 422-64-0
EINECS: 207-021-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR