Pentafluorobenzene ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
09-5450-5G
09-5450-25G
100197-352EA
70.77
USD
100197-352
100208-780
Pentafluorobenzene ≥99%
Pentafluorobenzene
Organo Fluorine
Formula:
C₆HF₅ Boiling Pt: 85 °C Melting Pt: –48 °C Density: 1,514 Flash Pt: 57 °F Storage Temperature: Ambient |
MDL Number:
MFCD00000286 CAS Number: 363-72-4 EINECS: 206-658-7 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...