Hexafluorobenzene ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Perfluorobenzene

09-4630-25G 09-4630-5G
100202-620EA 123.14 USD
100202-620 100207-542
Hexafluorobenzene ≥99%
Hexafluorobenzene

Organo Fluorine

Formula: C₆F₆
MW: 186.06 g/mol
Boiling Pt: 81…82 °C
Melting Pt: 4 °C
Density: 1.612
Flash Pt: 50 °F
Storage Temperature: Ambient
MDL Number: MFCD00000288
CAS Number: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,II
Merck Index: 13,04705

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR