meso-Tetraphenylporphine ≥97% (contains 1-3% chlorin)
Supplier: STREM CHEMICALS INC MS
Synonyms:
5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP
07-2160-2G
07-2160-10G
100208-276EA
99.08
USD
100208-276
100200-832
meso-Tetraphenylporphine ≥97% (contains 1-3% chlorin)
meso-Tetraphenylporphine
Porphine
Formula:
C₄₄H₃₀N₄ MW: 614.75 g/mol Density: 1,276 g/cm³ (20 °C) |
MDL Number:
MFCD00011680 CAS Number: 917-23-7 EINECS: 213-025-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...