Phthalocyanine
Supplier: STREM CHEMICALS INC MS
Synonyms:
Pigment Blue 16, 29H,31H-Phthalocyanine
07-1700-5G
07-1700-1G
07-1700-25G
100203-902EA
101.91
USD
100203-902
100195-984
100202-872
Phthalocyanine
Phthalocyanine
Formula:
C₃₂H₁₈N₈ MW: 514.55 g/mol Melting Pt: 210 °C Density: 1,447 g/cm³ (–113 °C) |
MDL Number:
MFCD00005085 CAS Number: 574-93-6 EINECS: 209-378-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...