2,2'-Bipyridyl ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
07-0290-25G
07-0290-100G
100203-896EA
138.71
USD
100203-896
100207-524
2,2'-Bipyridyl ≥99%
2,2'-Bipyridyl
Formula:
C₁₀H₈N₂ Boiling Pt: 273 °C Melting Pt: 69…70 °C Density: 1.326 g/cm³ (–150 °C) Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 EINECS: 206-674-4 UN: 2811 ADR: 6.1,III Merck Index: 13,03382 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...