1,3,5,7-Cyclooctatetraene ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cyclooctatetraene, COT
06-0350-1G
06-0350-5G
100209-506EA
70.77
USD
100209-506
100205-814
1,3,5,7-Cyclooctatetraene ≥98%
1,3,5,7-Cyclooctatetraene
Formula:
C₈H₈ Boiling Pt: 142…143 °C Melting Pt: –5 °C Density: 0,943 Flash Pt: 73 °F Storage Temperature: Refrigerator |
MDL Number:
MFCD00004161 CAS Number: 629-20-9 EINECS: 211-080-3 UN: 2358 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...