Lanthanum ≥99.9%, ingot
Supplier: STREM CHEMICALS INC MS
|
Danger
|
57-5747-100G
57-5747-25G
100204-090EA
642.58
USD
100204-090
100198-228
Lanthanum ≥99.9%, ingot
Lanthanum
Formula:
La Boiling Pt: 3454 °C Melting Pt: 920 °C Density: 6,166 Storage Temperature: Ambient |
MDL Number:
MFCD00011066 CAS Number: 7439-91-0 EINECS: 231-099-0 UN: 3208 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...