Barium carbonate ≥99.9% (Ba basis) (Sr < .03%)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
56-1020-500G
56-1020-100G
100204-632EA
360.92
USD
100204-632
100208-450
Barium carbonate ≥99.9% (Ba basis) (Sr < .03%)
Barium carbonate
Formula:
BaCO₃ Melting Pt: 1350 °C Density: 4.43 Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III Merck Index: 13,00972 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...