Antimony(V) chloride ≥99.999% (Sb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

51-2200-5G 51-2200-25G
100210-652EA 266.09 USD
100210-652 100205-962
Antimony(V) chloride ≥99.999% (Sb basis), PURATREM
Antimony (V) chloride
Formula: Cl₅Sb
Boiling Pt: 79 °C (22 mmHg)
Melting Pt: 2.8 °C
Density: 2.336
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8,II
Merck Index: 13,00701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR