Silver hexafluoroarsenate ≥98.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Silver hexafluoroarsenate(V)
47-0900-2G
47-0900-10G
100199-420EA
280.25
USD
100199-420
100205-958
Silver hexafluoroarsenate ≥98.5%
Silver hexafluoroarsenate
Formula:
AgAsF₆ Melting Pt: Dec. |
MDL Number:
MFCD00014142 CAS Number: 12005-82-2 UN: 1557 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...