Silver ≥99.99%, shot
Supplier: STREM CHEMICALS INC MS
47-0085-50G
47-0085-10G
100198-850EA
379.32
USD
100198-850
100198-848
Silver ≥99.99%, shot
Silver
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960.5 °C Density: 10.49 Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...