Palladium ≥99.9%, wire, Length 10 cm, Diameter 1.0 mm

Supplier: STREM CHEMICALS INC MS
Warning

46-2125-50CM 46-2125-10CM
100211-122EA 696.37 USD
100211-122 100211-120
Palladium ≥99.9%, wire, Length 10 cm, Diameter 1.0 mm
Palladium
Formula: Pd
Boiling Pt: 3140 °C
Melting Pt: 1552 °C
Density: 12.02
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR