Palladium(II) trifluoroacetate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

46-0280-1G 46-0280-5G
100200-474EA 195.32 USD
100200-474 100204-070
Palladium(II) trifluoroacetate ≥97%
Palladium (II) trifluoroacetate
Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR