Rhodium ≥99.8%, Powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-1870-2G
45-1870-500MG
100197-154EA
5645.97
USD
100197-154
100199-930
Rhodium ≥99.8%, Powder
Rhodium
Formula:
Rh MW: 102.9 g/mol Boiling Pt: 3727 °C Melting Pt: 1966 °C Density: 12,41 |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 EINECS: 231-125-0 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...