Sodium molybdate(VI) dihydrate ≥99.5% ACS

Supplier: STREM CHEMICALS INC MS
Warning

42-4234-500G 42-4234-100G
100207-678EA 300.06 USD
100207-678 100209-936
Sodium molybdate(VI) dihydrate ≥99.5% ACS
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR