Niobium ≥99.8%, rods

Supplier: STREM CHEMICALS INC MS
Warning

41-0070-25CM 41-0070-100CM 41-0070-5CM
100199-880EA 308.55 USD
100199-880 100197-508 100200-458
Niobium ≥99.8%, rods
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR