Niobium ≥99.8%, foil
Supplier: STREM CHEMICALS INC MS
41-0060-5PC
41-0060-1PC
100200-964EA
454.34
USD
100200-964
100207-672
Niobium ≥99.8%, foil
Niobium
Formula:
Nb MW: 92.91 g/mol Boiling Pt: 3700 Melting Pt: 2468 Density: 8.57 |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...