Strontium carbonate ≥99.995%, PURATREM

Supplier: STREM CHEMICALS INC MS

38-1100-50G 38-1100-10G
100211-076EA 401.97 USD
100211-076 100211-074
Strontium carbonate ≥99.995%, PURATREM
Strontium carbonate
Formula: CO₃Sr
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1497 °C
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR