Diethyl diselenide ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
34-0380-10G
34-0380-50G
100203-102EA
488.31
USD
100203-102
100199-234
Diethyl diselenide ≥97%
Diethyl diselenide
Metal Alkyls, Volatile Precursors for CVD
Formula:
C₄H₁₀Se₂ Boiling Pt: 85 °C (21 mmHg) |
MDL Number:
MFCD00015491 CAS Number: 628-39-7 EINECS: 211-042-6 UN: 3440 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...