Arsenic (III) sulfide ≥99.999% (As basis), fused lump, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Arsenic trisulphide, Arsenic trisulfide, Disrsenic trisulfide, Disrsenic trisulphide

33-1199-10G 33-1199-50G
100207-384EA 250.52 USD
100207-384 100196-942
Arsenic (III) sulfide ≥99.999% (As basis), fused lump, PURATREM
Arsenic (III) sulphide
Formula: As₂S₃
Boiling Pt: 707 °C
Melting Pt: 300 °C
Density: 3.43
MDL Number: MFCD00003435
CAS Number: 1303-33-9
EINECS: 215-117-4
UN: 1557
ADR: 6.1,II
Merck Index: 12,00846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR