Zinc ≥99.9999%, wire
Supplier: STREM CHEMICALS INC MS
30-0090-1MT
30-0090-5MT
100199-382EA
226.46
USD
100199-382
100199-384
Zinc ≥99.9999%, wire
Zinc
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...