Zinc ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

30-0040-1PC 30-0040-5PC
100211-066EA 104.74 USD
100211-066 100211-068
Zinc ≥99.9%, foil
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR