Copper(I) chloride ≥99.99% (Cu basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cuprous chloride, Copper chloride

29-0360-50G 29-0360-10G
100206-562EA 413.29 USD
100206-562 100206-560
Copper(I) chloride ≥99.99% (Cu basis), PURATREM
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR