Copper ≥99.999%, rods

Supplier: STREM CHEMICALS INC MS

29-0080-500G 29-0080-25G 29-0080-100G
100196-874EA 2169.78 USD
100196-874 100208-876 100208-016
Copper ≥99.999%, rods
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR