Nickel(II) nitrate hexahydrate ≥99.9985% (Ni basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

28-1440-100G 28-1440-25G
100200-264EA 222.22 USD
100200-264 100205-074
Nickel(II) nitrate hexahydrate ≥99.9985% (Ni basis), PURATREM
Nickel (II) nitrate hexahydrate
Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: 56.7 °C
Density: 2.05
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III
Merck Index: 12,06596

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR