Nickel ≥99%, foil

Supplier: STREM CHEMICALS INC MS
Warning

28-0001-100CM 28-0001-20CM
100199-848EA 447.26 USD
100199-848 100206-550
Nickel ≥99%, foil
Nickel
Formula: Ni
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.90
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR